Assay Details
| Gene Symbol: CG6845 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144386.3 | NP_001137858.1 | 4 - 5 | 904 | 106 | |
| NM_001144387.3 | NP_001137859.1 | 6 - 7 | 2129 | 106 | ||
| NM_138156.5 | NP_612000.3 | 8 - 9 | 2011 | 106 | ||
| GenBank mRNA | AY051619.1 | - | 6 - 7 | 1406 | 106 | |
| BT003264.1 | - | 6 - 7 | 2109 | 106 | ||
More Information
Set Membership: |
|

CG6845
Genomic Map