Assay Details
| Gene Symbol: Vdup1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169832.2 | NP_001163303.1 | 3 - 4 | 1241 | 128 | |
| NM_001274255.1 | NP_001261184.1 | 3 - 4 | 801 | 128 | ||
| NM_138160.4 | NP_612004.2 | 3 - 4 | 809 | 128 | ||
| NM_167798.3 | NP_728479.1 | 4 - 5 | 879 | 128 | ||
| NM_206221.3 | NP_995943.1 | 4 - 5 | 1093 | 128 | ||
| GenBank mRNA | BT150163.1 | - | 4 - 5 | 873 | 128 | |
| BT150199.1 | - | 3 - 4 | 1235 | 128 | ||
More Information
Set Membership: |
|

Vdup1
Genomic Map