Assay Details
| Gene Symbol: Pxn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079167.5 | NP_523891.2 | 2 - 3 | 538 | 59 | |
| NM_167957.3 | NP_728759.1 | 2 - 3 | 538 | 59 | ||
| NM_206253.3 | NP_995975.1 | 1 - 2 | 232 | 59 | ||
| NM_206254.3 | NP_995976.1 | 2 - 3 | 608 | 59 | ||
| NM_206255.3 | NP_995977.1 | 2 - 3 | 596 | 59 | ||
| GenBank mRNA | AY052120.1 | - | 2 - 3 | 546 | 59 | |
| BT120118.1 | - | 2 - 3 | 524 | 59 | ||
More Information
Set Membership: |
|

Pxn
Genomic Map