Assay Details
| Gene Symbol: PAN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104016.2 | NP_001097486.1 | 11 - 12 | 2245 | 58 | |
| NM_001104017.2 | NP_001097487.1 | 12 - 13 | 2702 | 58 | ||
| NM_001104018.2 | NP_001097488.1 | 13 - 14 | 2834 | 58 | ||
| NM_139510.4 | NP_647767.1 | 14 - 15 | 2838 | 58 | ||
| NM_167975.3 | NP_728797.1 | 13 - 14 | 2690 | 58 | ||
| NM_167976.3 | NP_728798.1 | 13 - 14 | 2677 | 58 | ||
| NM_167977.3 | NP_728799.1 | 13 - 14 | 2795 | 58 | ||
| NM_167978.3 | NP_728800.1 | 13 - 14 | 2760 | 58 | ||
| NM_167979.3 | NP_728801.1 | 12 - 13 | 2584 | 58 | ||
| NM_167980.3 | NP_728802.2 | 12 - 13 | 2338 | 58 | ||
| NM_167981.2 | NP_728803.2 | 11 - 12 | 2401 | 58 | ||
| NM_167982.3 | NP_728804.1 | 11 - 12 | 2379 | 58 | ||
| NM_167983.3 | NP_728805.1 | 11 - 12 | 2388 | 58 | ||
| NM_167984.3 | NP_728806.1 | 11 - 12 | 2322 | 58 | ||
| NM_206260.3 | NP_995982.1 | 14 - 15 | 2728 | 58 | ||
| NM_206261.3 | NP_995983.1 | 13 - 14 | 2785 | 58 | ||
| GenBank mRNA | AY061183.1 | - | 14 - 15 | 2796 | 58 | |
| BT100048.1 | - | 11 - 12 | 2366 | 58 | ||
| BT125980.1 | - | 13 - 14 | 2651 | 58 | ||
More Information
Set Membership: |
|

PAN3
Genomic Map