Assay Details
| Gene Symbol: ckd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139539.2 | NP_647796.2 | 4 - 5 | 787 | 57 | |
| NM_168009.2 | NP_728847.2 | 3 - 4 | 549 | 57 | ||
| NM_206264.2 | NP_995986.1 | 3 - 4 | 982 | 57 | ||
| GenBank mRNA | BT011465.1 | - | 3 - 4 | 981 | 57 | |
More Information
Set Membership: |
|

ckd
Genomic Map