Assay Details
| Gene Symbol: PIG-B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139575.3 | NP_647832.1 | 2 - 3 | 354 | 66 | |
| NM_206269.3 | NP_995991.1 | 2 - 3 | 354 | 66 | ||
| GenBank mRNA | BT003240.1 | - | 2 - 3 | 336 | 66 | |
More Information
Set Membership: |
|

PIG-B
Genomic Map