Assay Details
Gene Symbol: enc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104026.2 | NP_001097496.1 | 7 - 8 | 2407 | 54 | |
NM_001274449.1 | NP_001261378.1 | 7 - 8 | 2350 | 54 | ||
NM_080026.3 | NP_524765.2 | 7 - 8 | 2350 | 54 | ||
NM_206270.2 | NP_995992.1 | 7 - 8 | 2365 | 54 | ||
GenBank mRNA | BT050449.1 | - | 7 - 8 | 2348 | 54 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
