Assay Details
| Gene Symbol: Rdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_206271.2 | NP_995993.1 | 1 | 115 | 60 | |
| GenBank mRNA | BT100092.4 | - | 1 | 40 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rdh| Gene Symbol: Rdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_206271.2 | NP_995993.1 | 1 | 115 | 60 | |
| GenBank mRNA | BT100092.4 | - | 1 | 40 | 60 | |
Set Membership: |
|