Assay Details
| Gene Symbol: Clk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014576.2 | NP_001014576.1 | 3 - 4 | 571 | 76 | |
| NM_079240.3 | NP_523964.2 | 3 - 4 | 559 | 76 | ||
More Information
Set Membership: |
|

Clk
Genomic Map