Assay Details
| Gene Symbol: Clk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014574.2 | NP_001014574.1 | 5 - 6 | 966 | 68 | |
| NM_001014576.2 | NP_001014576.1 | 5 - 6 | 968 | 68 | ||
| NM_079240.3 | NP_523964.2 | 5 - 6 | 956 | 68 | ||
| NM_170628.2 | NP_729294.2 | 2 - 3 | 299 | 68 | ||
| NM_206299.2 | NP_996021.2 | 2 - 3 | 299 | 68 | ||
More Information
Set Membership: |
|

Clk
Genomic Map