Assay Details
| Gene Symbol: PGRP-LA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_206304.2 | NP_996026.1 | 2 - 3 | 420 | 57 | |
| NM_206305.2 | NP_996027.1 | 2 - 3 | 420 | 57 | ||
| GenBank mRNA | AY060314.1 | - | 2 - 3 | 419 | 57 | |
More Information
Set Membership: |
|

PGRP-LA
Genomic Map