Assay Details
| Gene Symbol: PGRP-LA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274694.1 | NP_001261623.1 | 2 - 3 | 290 | 84 | |
| NM_206304.2 | NP_996026.1 | 3 - 4 | 643 | 84 | ||
| NM_206307.2 | NP_996029.1 | 2 - 3 | 551 | 84 | ||
| GenBank mRNA | AF207535.1 | - | 2 - 3 | 523 | 84 | |
| BT001411.1 | - | 2 - 3 | 522 | 84 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001274694.1 transcripts.
Set Membership: |
|

PGRP-LA
Genomic Map