Assay Details
| Gene Symbol: PGRP-LA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274694.1 | NP_001261623.1 | 4 - 5 | 730 | 84 | |
| NM_206304.2 | NP_996026.1 | 5 - 6 | 1083 | 84 | ||
| NM_206305.2 | NP_996027.1 | 5 - 6 | 1071 | 84 | ||
| NM_206306.2 | NP_996028.1 | 4 - 5 | 978 | 84 | ||
| NM_206307.2 | NP_996029.1 | 4 - 5 | 991 | 84 | ||
| GenBank mRNA | AF207535.1 | - | 4 - 5 | 963 | 84 | |
| AF207536.1 | - | 4 - 5 | 979 | 84 | ||
| AY060314.1 | - | 5 - 6 | 1070 | 84 | ||
| BT001411.1 | - | 4 - 5 | 962 | 84 | ||
More Information
Set Membership: |
|

PGRP-LA
Genomic Map