Assay Details
| Gene Symbol: CG43689 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272298.2 | NP_001259227.1 | 10 - 11 | 4869 | 80 | |
| NM_001272300.2 | NP_001259229.1 | 9 - 10 | 4539 | 80 | ||
| NM_001272301.2 | NP_001259230.1 | 10 - 11 | 4713 | 80 | ||
| NM_001297921.1 | NP_001284850.1 | 10 - 11 | 4449 | 80 | ||
| NM_001370021.1 | NP_001356932.1 | 9 - 10 | 4774 | 80 | ||
More Information
Set Membership: |
|

CG43689
Genomic Map