Assay Details
| Gene Symbol: Tip60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272304.1 | NP_001259233.1 | 4 - 5 | 2245 | 82 | |
| NM_001272305.1 | NP_001259234.1 | 4 - 5 | 1729 | 82 | ||
| NM_131923.4 | NP_572151.1 | 4 - 5 | 2267 | 82 | ||
| GenBank mRNA | AY051795.1 | - | 4 - 5 | 2222 | 82 | |
More Information
Set Membership: |
|

Tip60
Genomic Map