Assay Details
| Gene Symbol: Proc-R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014723.2 | NP_001014723.1 | 8 - 9 | 1888 | 100 | |
| NM_001014724.2 | NP_001014724.1 | 8 - 9 | 1555 | 100 | ||
| NM_001297939.1 | NP_001284868.1 | 7 - 8 | 1667 | 100 | ||
| NM_131955.2 | NP_572183.1 | 8 - 9 | 1649 | 100 | ||
| NM_167020.2 | NP_726950.1 | 8 - 9 | 1844 | 100 | ||
| GenBank mRNA | AY128481.1 | - | 8 - 9 | 1647 | 100 | |
More Information
Set Membership: |
|

Proc-R
Genomic Map