Assay Details
| Gene Symbol: CG32772 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001297941.1 | NP_001284870.1 | 2 - 3 | 566 | 72 | |
| NM_131968.5 | NP_572196.3 | 1 - 2 | 698 | 72 | ||
| GenBank mRNA | BT022160.2 | - | 1 - 2 | 652 | 72 | |
More Information
Set Membership: |
|

CG32772
Genomic Map