Assay Details
| Gene Symbol: CG32772 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001297941.1 | NP_001284870.1 | 3 - 4 | 893 | 82 | |
| NM_131968.5 | NP_572196.3 | 2 - 3 | 1025 | 82 | ||
| GenBank mRNA | BT022160.2 | - | 2 - 3 | 979 | 82 | |
More Information
Set Membership: |
|

CG32772
Genomic Map