Assay Details
| Gene Symbol: CG32772 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103409.2 | NP_001096879.2 | 4 - 5 | 1033 | 74 | |
| NM_001297941.1 | NP_001284870.1 | 6 - 7 | 1793 | 74 | ||
| NM_001297942.1 | NP_001284871.1 | 4 - 5 | 1033 | 74 | ||
| NM_131968.5 | NP_572196.3 | 5 - 6 | 1925 | 74 | ||
| GenBank mRNA | BT022160.2 | - | 5 - 6 | 1879 | 74 | |
More Information
Set Membership: |
|

CG32772
Genomic Map