Assay Details
| Gene Symbol: Usp16-45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001297961.1 | NP_001284890.1 | 4 - 5 | 2380 | 87 | |
| NM_131992.3 | NP_572220.1 | 4 - 5 | 2380 | 87 | ||
| NM_167035.2 | NP_726993.1 | 4 - 5 | 2305 | 87 | ||
| NM_167036.2 | NP_726994.1 | 4 - 5 | 2332 | 87 | ||
| NM_167037.2 | NP_726995.1 | 4 - 5 | 2356 | 87 | ||
| GenBank mRNA | AY051846.1 | - | 4 - 5 | 2442 | 87 | |
More Information
Set Membership: |
|

Usp16-45
Genomic Map