Assay Details
| Gene Symbol: frma | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131998.3 | NP_572226.2 | 1 - 2 | 216 | 94 | |
| NM_167038.2 | NP_726999.2 | 1 - 2 | 216 | 94 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

frma| Gene Symbol: frma | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131998.3 | NP_572226.2 | 1 - 2 | 216 | 94 | |
| NM_167038.2 | NP_726999.2 | 1 - 2 | 216 | 94 | ||
Set Membership: |
|