Assay Details
| Gene Symbol: CG42699 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_132035.3 | NP_572263.2 | 2 - 3 | 1709 | 85 | |
| GenBank mRNA | BT030997.1 | - | 2 - 3 | 1687 | 85 | |
More Information
Set Membership: |
|

CG42699
Genomic Map