Assay Details
| Gene Symbol: CG42699 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_132035.3 | NP_572263.2 | 3 - 4 | 1915 | 100 | |
| GenBank mRNA | BT030997.1 | - | 3 - 4 | 1893 | 100 | |
More Information
Set Membership: |
|

CG42699
Genomic Map