Assay Details
| Gene Symbol: CG5937 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103418.3 | NP_001096888.2 | 5 - 6 | 1428 | 68 | |
| NM_132061.3 | NP_572289.2 | 5 - 6 | 1446 | 68 | ||
More Information
Set Membership: |
|

CG5937
Genomic Map