Assay Details
| Gene Symbol: CG5937 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103418.3 | NP_001096888.2 | 8 - 9 | 3521 | 82 | |
| NM_132061.3 | NP_572289.2 | 8 - 9 | 3539 | 82 | ||
More Information
Set Membership: |
|

CG5937
Genomic Map