Assay Details
| Gene Symbol: Ca-alpha1T | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103419.2 | NP_001096889.1 | 14 - 15 | 3409 | 88 | |
| NM_001258615.1 | NP_001245544.1 | 13 - 14 | 3385 | 88 | ||
| NM_001258616.1 | NP_001245545.1 | 14 - 15 | 3394 | 88 | ||
| NM_001272338.1 | NP_001259267.1 | 15 - 16 | 3448 | 88 | ||
| NM_132068.3 | NP_572296.2 | 14 - 15 | 3409 | 88 | ||
More Information
Set Membership: |
|

Ca-alpha1T
Genomic Map