Assay Details
| Gene Symbol: CadN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042903.4 | NP_001036368.2 | 12 - 13 | 5393 | 77 | |
| NM_136011.4 | NP_609855.3 | 13 - 14 | 6087 | 77 | ||
| GenBank mRNA | BT132666.1 | - | 12 - 13 | 5360 | 77 | |
More Information
Set Membership: |
|

CadN2
Genomic Map