Assay Details
| Gene Symbol: CG5790 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299108.1 | NP_001286037.1 | 1 | 2377 | 122 | |
| NM_136032.4 | NP_609876.2 | 1 | 2377 | 122 | ||
| GenBank mRNA | BT133199.1 | - | 1 | 2294 | 122 | |
More Information
Set Membership: |
|

CG5790
Genomic Map