Assay Details
| Gene Symbol: CG13551 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144045.2 | NP_652302.1 | 2 - 3 | 137 | 61 | |
| NM_166597.2 | NP_726315.1 | 2 - 3 | 182 | 61 | ||
| GenBank mRNA | BT030768.1 | - | 2 - 3 | 134 | 61 | |
More Information
Set Membership: |
|

CG13551
Genomic Map