Assay Details
| Gene Symbol: Sytbeta | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144170.2 | NP_652427.2 | 2 | 574 | 60 | |
| GenBank mRNA | BT133101.1 | - | 2 | 575 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sytbeta| Gene Symbol: Sytbeta | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144170.2 | NP_652427.2 | 2 | 574 | 60 | |
| GenBank mRNA | BT133101.1 | - | 2 | 575 | 60 | |
Set Membership: |
|