Assay Details
| Gene Symbol: RecQ4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144350.2 | NP_652607.1 | 4 - 5 | 4306 | 76 | |
| GenBank mRNA | BT150480.1 | - | 4 - 5 | 4226 | 76 | |
More Information
Set Membership: |
|

RecQ4
Genomic Map