Assay Details
| Gene Symbol: SC35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001201865.2 | NP_001188794.1 | 1 - 2 | 206 | 60 | |
| NM_001259076.2 | NP_001246005.1 | 1 - 2 | 206 | 60 | ||
| NM_144355.4 | NP_652612.1 | 1 - 2 | 206 | 60 | ||
| GenBank mRNA | AF232775.1 | - | 1 - 2 | 144 | 60 | |
| AY069584.1 | - | 1 - 2 | 144 | 60 | ||
More Information
Set Membership: |
|

SC35
Genomic Map