Assay Details
| Gene Symbol: SC35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001201865.2 | NP_001188794.1 | 2 - 3 | 335 | 55 | |
| NM_001259076.2 | NP_001246005.1 | 2 - 3 | 335 | 55 | ||
| NM_144355.4 | NP_652612.1 | 2 - 3 | 335 | 55 | ||
| NM_205967.2 | NP_995689.1 | 2 - 3 | 238 | 55 | ||
| GenBank mRNA | AF232775.1 | - | 2 - 3 | 273 | 55 | |
| AY069584.1 | - | 2 - 3 | 273 | 55 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_205967.2 transcripts.
Set Membership: |
|

SC35
Genomic Map