Assay Details
| Gene Symbol: Ugt37D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299060.1 | NP_001285989.1 | 1 - 2 | 753 | 76 | |
| NM_144370.3 | NP_652627.2 | 2 - 3 | 949 | 76 | ||
| NM_165193.2 | NP_724013.1 | 2 - 3 | 969 | 76 | ||
| GenBank mRNA | BT056249.1 | - | 2 - 3 | 945 | 76 | |
More Information
Set Membership: |
|

Ugt37D1
Genomic Map