Assay Details
| Gene Symbol: lectin-24A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144380.2 | NP_652637.1 | 1 | 836 | 84 | |
| GenBank mRNA | BT100093.4 | - | 1 | 791 | 84 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

lectin-24A| Gene Symbol: lectin-24A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144380.2 | NP_652637.1 | 1 | 836 | 84 | |
| GenBank mRNA | BT100093.4 | - | 1 | 791 | 84 | |
Set Membership: |
|