Assay Details
| Gene Symbol: lectin-29Ca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144383.2 | NP_652640.1 | 1 | 673 | 134 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

lectin-29Ca| Gene Symbol: lectin-29Ca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144383.2 | NP_652640.1 | 1 | 673 | 134 | |
Set Membership: |
|