Assay Details
| Gene Symbol: lectin-24Db | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144384.3 | NP_652641.1 | 1 | 1061 | 70 | |
| GenBank mRNA | AY061025.1 | - | 1 | 1061 | 70 | |
| AY119000.1 | - | 1 | 1048 | 70 | ||
More Information
Set Membership: |
|

lectin-24Db
Genomic Map