Assay Details
| Gene Symbol: lectin-21Ca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144385.3 | NP_652642.2 | 1 | 797 | 66 | |
| GenBank mRNA | BT032781.1 | - | 1 | 797 | 66 | |
| BT032802.1 | - | 1 | 797 | 66 | ||
More Information
Set Membership: |
|

lectin-21Ca
Genomic Map