Assay Details
| Gene Symbol: Cpr65Ax2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274538.1 | NP_001261467.1 | 1 - 2 | 161 | 79 | |
| NM_144403.3 | NP_652660.1 | 1 - 2 | 161 | 79 | ||
| GenBank mRNA | BT120012.1 | - | 1 - 2 | 114 | 79 | |
| Gene Symbol: Cpr65Ax1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104053.3 | NP_001097523.1 | 1 - 2 | 162 | 79 | |
More Information
Set Membership: |
|

Cpr65Ax2
Genomic Map