Assay Details
| Gene Symbol: CG18814 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144416.2 | NP_652673.2 | 1 - 2 | 147 | 89 | |
| Gene Symbol: Mdh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BT029274.1 | - | 1474 | 89 | ||
More Information
Set Membership: |
|

CG18814
Genomic Map