Assay Details
| Gene Symbol: shot | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144185.2 | NP_001137657.1 | 13 - 14 | 4477 | 71 | |
| NM_001144186.2 | NP_001137658.1 | 13 - 14 | 4477 | 71 | ||
| NM_001144187.2 | NP_001137659.1 | 13 - 14 | 4477 | 71 | ||
| NM_001144188.2 | NP_001137660.1 | 13 - 14 | 4700 | 71 | ||
| NM_001144189.2 | NP_001137661.1 | 13 - 14 | 4700 | 71 | ||
| NM_001169677.1 | NP_001163148.1 | 13 - 14 | 4477 | 71 | ||
| NM_001169678.1 | NP_001163149.1 | 13 - 14 | 4700 | 71 | ||
| NM_001169679.2 | NP_001163150.1 | 10 - 11 | 4493 | 71 | ||
| NM_001259378.1 | NP_001246307.1 | 10 - 11 | 4493 | 71 | ||
| NM_001259379.1 | NP_001246308.1 | 10 - 11 | 4493 | 71 | ||
| NM_001259380.1 | NP_001246309.1 | 14 - 15 | 4495 | 71 | ||
| NM_001259381.1 | NP_001246310.1 | 14 - 15 | 4537 | 71 | ||
| NM_001259383.1 | NP_001246312.1 | 10 - 11 | 4493 | 71 | ||
| NM_001259384.1 | NP_001246313.1 | 10 - 11 | 4493 | 71 | ||
| NM_079009.3 | NP_523733.2 | 13 - 14 | 4700 | 71 | ||
| NM_166015.2 | NP_725335.1 | 13 - 14 | 4477 | 71 | ||
| NM_166016.2 | NP_725336.1 | 13 - 14 | 4477 | 71 | ||
| NM_166019.3 | NP_725339.1 | 10 - 11 | 4493 | 71 | ||
More Information
Set Membership: |
|

shot
Genomic Map