Assay Details
| Gene Symbol: shot | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144186.2 | NP_001137658.1 | 20 - 21 | 15057 | 63 | |
| NM_001144189.2 | NP_001137661.1 | 20 - 21 | 15280 | 63 | ||
| NM_001169676.1 | NP_001163147.1 | 13 - 14 | 14173 | 63 | ||
| NM_079009.3 | NP_523733.2 | 20 - 21 | 15280 | 63 | ||
| NM_166015.2 | NP_725335.1 | 20 - 21 | 15057 | 63 | ||
| NM_166016.2 | NP_725336.1 | 20 - 21 | 15057 | 63 | ||
| NM_166017.2 | NP_725337.1 | 16 - 17 | 14380 | 63 | ||
| NM_166018.3 | NP_725338.1 | 13 - 14 | 14173 | 63 | ||
| NM_166019.3 | NP_725339.1 | 17 - 18 | 25108 | 63 | ||
More Information
Set Membership: |
|

shot
Genomic Map