Assay Details
| Gene Symbol: shot | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144186.2 | NP_001137658.1 | 21 - 22 | 15100 | 69 | |
| NM_001144189.2 | NP_001137661.1 | 21 - 22 | 15323 | 69 | ||
| NM_001169676.1 | NP_001163147.1 | 14 - 15 | 14216 | 69 | ||
| NM_001169677.1 | NP_001163148.1 | 22 - 23 | 15160 | 69 | ||
| NM_001169678.1 | NP_001163149.1 | 22 - 23 | 15383 | 69 | ||
| NM_001169679.2 | NP_001163150.1 | 19 - 20 | 15176 | 69 | ||
| NM_001259379.1 | NP_001246308.1 | 19 - 20 | 15176 | 69 | ||
| NM_001259380.1 | NP_001246309.1 | 23 - 24 | 15178 | 69 | ||
| NM_001259381.1 | NP_001246310.1 | 23 - 24 | 15220 | 69 | ||
| NM_001259382.1 | NP_001246311.1 | 11 - 12 | 11036 | 69 | ||
| NM_001259383.1 | NP_001246312.1 | 19 - 20 | 15188 | 69 | ||
| NM_079009.3 | NP_523733.2 | 21 - 22 | 15323 | 69 | ||
| NM_166015.2 | NP_725335.1 | 21 - 22 | 15100 | 69 | ||
| NM_166016.2 | NP_725336.1 | 21 - 22 | 15100 | 69 | ||
| NM_166017.2 | NP_725337.1 | 17 - 18 | 14423 | 69 | ||
| NM_166018.3 | NP_725338.1 | 14 - 15 | 14216 | 69 | ||
| NM_166019.3 | NP_725339.1 | 18 - 19 | 25151 | 69 | ||
More Information
Set Membership: |
|

shot
Genomic Map