Assay Details
| Gene Symbol: shot | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144187.2 | NP_001137659.1 | 25 - 26 | 15776 | 86 | |
| NM_001169676.1 | NP_001163147.1 | 19 - 20 | 14937 | 86 | ||
| NM_166015.2 | NP_725335.1 | 26 - 27 | 15821 | 86 | ||
More Information
Set Membership: |
|

shot
Genomic Map