Assay Details
| Gene Symbol: shot | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144185.2 | NP_001137657.1 | 28 - 29 | 16423 | 80 | |
| NM_001144187.2 | NP_001137659.1 | 28 - 29 | 16408 | 80 | ||
| NM_001144188.2 | NP_001137660.1 | 28 - 29 | 16646 | 80 | ||
| NM_001169676.1 | NP_001163147.1 | 22 - 23 | 15569 | 80 | ||
| NM_001259379.1 | NP_001246308.1 | 27 - 28 | 16544 | 80 | ||
| NM_079009.3 | NP_523733.2 | 29 - 30 | 16691 | 80 | ||
| NM_166015.2 | NP_725335.1 | 29 - 30 | 16453 | 80 | ||
| NM_166016.2 | NP_725336.1 | 29 - 30 | 16468 | 80 | ||
| NM_166017.2 | NP_725337.1 | 25 - 26 | 15791 | 80 | ||
| NM_166018.3 | NP_725338.1 | 22 - 23 | 15584 | 80 | ||
| NM_166019.3 | NP_725339.1 | 26 - 27 | 26519 | 80 | ||
More Information
Set Membership: |
|

shot
Genomic Map