Assay Details
| Gene Symbol: shot | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144185.2 | NP_001137657.1 | 26 - 27 | 15964 | 94 | |
| NM_001144186.2 | NP_001137658.1 | 27 - 28 | 16009 | 94 | ||
| NM_001144188.2 | NP_001137660.1 | 26 - 27 | 16187 | 94 | ||
| NM_001144189.2 | NP_001137661.1 | 27 - 28 | 16232 | 94 | ||
| NM_001169677.1 | NP_001163148.1 | 28 - 29 | 16069 | 94 | ||
| NM_001169678.1 | NP_001163149.1 | 28 - 29 | 16292 | 94 | ||
| NM_001169679.2 | NP_001163150.1 | 25 - 26 | 16085 | 94 | ||
| NM_001259378.1 | NP_001246307.1 | 24 - 25 | 16013 | 94 | ||
| NM_001259379.1 | NP_001246308.1 | 25 - 26 | 16085 | 94 | ||
| NM_001259380.1 | NP_001246309.1 | 29 - 30 | 16087 | 94 | ||
| NM_001259381.1 | NP_001246310.1 | 29 - 30 | 16129 | 94 | ||
| NM_001259382.1 | NP_001246311.1 | 17 - 18 | 11945 | 94 | ||
| NM_001259383.1 | NP_001246312.1 | 25 - 26 | 16097 | 94 | ||
| NM_001259384.1 | NP_001246313.1 | 24 - 25 | 16013 | 94 | ||
| NM_079009.3 | NP_523733.2 | 27 - 28 | 16232 | 94 | ||
| NM_166016.2 | NP_725336.1 | 27 - 28 | 16009 | 94 | ||
| NM_166017.2 | NP_725337.1 | 23 - 24 | 15332 | 94 | ||
| NM_166018.3 | NP_725338.1 | 20 - 21 | 15125 | 94 | ||
| NM_166019.3 | NP_725339.1 | 24 - 25 | 26060 | 94 | ||
More Information
Set Membership: |
|

shot
Genomic Map