Assay Details
| Gene Symbol: CG8485 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_137098.3 | NP_610942.2 | 2 - 3 | 528 | 72 | |
| NM_166041.2 | NP_725370.1 | 2 - 3 | 320 | 72 | ||
| NM_166042.2 | NP_725371.1 | 1 - 2 | 505 | 72 | ||
More Information
Set Membership: |
|

CG8485
Genomic Map