Assay Details
| Gene Symbol: CG8485 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_137098.3 | NP_610942.2 | 7 - 8 | 1544 | 66 | |
| NM_166041.2 | NP_725370.1 | 7 - 8 | 1336 | 66 | ||
| NM_166042.2 | NP_725371.1 | 6 - 7 | 1521 | 66 | ||
More Information
Set Membership: |
|

CG8485
Genomic Map