Assay Details
| Gene Symbol: Obp50c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_166052.3 | NP_725387.3 | 3 - 4 | 1254 | 77 | |
| Gene Symbol: Obp50b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_166051.3 | NP_725386.1 | 3 - 4 | 1254 | 77 | |
| GenBank mRNA | BT088819.1 | - | 3 - 4 | 1254 | 77 | |
More Information
Set Membership: |
|

Obp50c
Genomic Map