Assay Details
| Gene Symbol: hui | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_137147.5 | NP_610991.1 | 2 - 3 | 141 | 91 | |
| NM_166071.4 | NP_725421.1 | 1 - 2 | 228 | 91 | ||
| GenBank mRNA | AY118599.1 | - | 1 - 2 | 198 | 91 | |
More Information
Set Membership: |
|

hui
Genomic Map